PRI424058489 Princeton BioMolecular Research, Inc.

N-(2,2-difluoroethyl)-N-[2-(3,5-dimethyl-1H-pyrazol-1-yl)propyl]amine

  • MDL number MFCD15976297
  • Cas Number : 1245807-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart