PRI424058558 Princeton BioMolecular Research, Inc.

3-(5-methyl-2H-pyrazolo[3,4-b]pyridin-2-yl)propanoic acid

  • MDL number MFCD15976342
  • Cas Number : 1245807-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart