PRI424058560 Princeton BioMolecular Research, Inc.

3-(trifluoromethyl)-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD15976344
  • Cas Number : 591234-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart