PRI424058589 Princeton BioMolecular Research, Inc.

1-(1,3-thiazol-2-ylsulfonyl)piperazine

  • MDL number MFCD15976367
  • Cas Number : 1245772-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart