PRI424058683 Princeton BioMolecular Research, Inc.

1-ethyl-3-(trifluoromethyl)-1H-pyrazole-5-sulfonyl chloride

  • MDL number MFCD15976424
  • Cas Number : 1245823-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart