PRI424058743 Princeton BioMolecular Research, Inc.

[1-(difluoromethyl)-1H-pyrazol-3-yl]acetonitrile

  • MDL number MFCD15976768
  • Cas Number : 1260659-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart