PRI424058783 Princeton BioMolecular Research, Inc.

ethyl 5,5-difluoro-2,4-dioxopentanoate

  • MDL number MFCD14657183
  • Cas Number : 1245773-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart