PRI424058799 Princeton BioMolecular Research, Inc.

methyl 1-methyl-5-nitro-1H-pyrazole-3-carboxylate

  • MDL number MFCD18262314
  • Cas Number : 796038-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart