PRI424058807 Princeton BioMolecular Research, Inc.

1-methyl-5-nitro-1H-pyrazole-3-carbonitrile

  • MDL number MFCD18262318
  • Cas Number : 1245772-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart