PRI424058823 Princeton BioMolecular Research, Inc.

3-(difluoromethyl)-4-methyl-1H-pyrazole

  • MDL number MFCD18262334
  • Cas Number : 1245772-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart