PRI424058886 Princeton BioMolecular Research, Inc.

2-[(1-ethyl-1H-pyrazol-5-yl)methoxy]aniline

  • MDL number MFCD18262373
  • Cas Number : 1245822-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart