PRI424058907 Princeton BioMolecular Research, Inc.

methyl 5-(chlorosulfonyl)-1-isopropyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD18262380
  • Cas Number : 1260658-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart