PRI424058908 Princeton BioMolecular Research, Inc.

methyl 5-(chlorosulfonyl)-1-ethyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD18262381
  • Cas Number : 1260659-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart