PRI424058915 Princeton BioMolecular Research, Inc.

methyl [3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]acetate

  • MDL number MFCD18262382
  • Cas Number : 1260379-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart