PRI424058951 Princeton BioMolecular Research, Inc.

(1-ethyl-1H-pyrazol-5-yl)acetonitrile

  • MDL number MFCD18262403
  • Cas Number : 1260659-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart