PRI424058955 Princeton BioMolecular Research, Inc.

[1-ethyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]acetonitrile

  • MDL number MFCD18262407
  • Cas Number : 1260659-32-2
  • Molecular Weight : 203.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart