PRI424058964 Princeton BioMolecular Research, Inc.

(1-methyl-3-nitro-1H-pyrazol-5-yl)acetic acid

  • MDL number MFCD18262415
  • Cas Number : 1260658-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart