PRI424058978 Princeton BioMolecular Research, Inc.

[1-(2,2-difluoroethyl)-1H-pyrazol-3-yl]acetic acid

  • MDL number MFCD18262427
  • Cas Number : 1260658-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart