PRI424058992 Princeton BioMolecular Research, Inc.

3-(chloromethyl)-1-ethyl-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD18262441
  • Cas Number : 1260659-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart