PRI424059016 Princeton BioMolecular Research, Inc.

4-[(1-ethyl-1H-pyrazol-5-yl)methoxy]aniline

  • MDL number MFCD19691701
  • Cas Number : 1262865-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart