PRI424059025 Princeton BioMolecular Research, Inc.

3-nitro-1H-pyrazole-4-carboxamide

  • MDL number MFCD16619866
  • Cas Number : 39205-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart