PRI424059044 Princeton BioMolecular Research, Inc.

[5-(hydroxymethyl)-1-methyl-pyrazol-4-yl]methanol

  • MDL number MFCD19691710
  • Cas Number : 1281984-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart