PRI424059087 Princeton BioMolecular Research, Inc.

1-methyl-5-nitro-pyrazole-3-carbaldehyde

  • MDL number MFCD20491491
  • Cas Number : 1215121-74-6
  • Molecular Weight : 155.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart