PRI424059161 Princeton BioMolecular Research, Inc.

2-(2,2-difluoroethyl)-5-nitro-pyrazole-3-carboxylic acid

  • MDL number MFCD20491543
  • Cas Number : 1328640-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart