PRI424059192 Princeton BioMolecular Research, Inc.

2-(2-cyclopentylpyrazol-3-yl)acetonitrile

  • MDL number MFCD20491572
  • Cas Number : 1328640-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart