PRI424059210 Princeton BioMolecular Research, Inc.

1-[1-(2,2-difluoroethyl)pyrazol-3-yl]-N-methyl-methanamine

  • MDL number MFCD20491583
  • Cas Number : 1345510-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart