PRI424059233 Princeton BioMolecular Research, Inc.

4-(3-Bromo-phenyl)-1-methyl-1H-pyrazole

  • MDL number MFCD19322917
  • Cas Number : 141938-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart