PRI424059235 Princeton BioMolecular Research, Inc.

4-(4-Bromo-phenyl)-1-isopropyl-1H-pyrazole

  • MDL number MFCD20491974
  • Cas Number : 1351386-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart