PRI424059236 Princeton BioMolecular Research, Inc.

2-Bromo-4-(difluoro-methanesulfonyl)-1-methyl-benzene

  • MDL number MFCD20492028
  • Molecular Weight : 285.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-[(difluoromethyl)sulfonyl]-1-methylbenzene
Inchi Code 1S/C8H7BrF2O2S/c1-5-2-3-6(4-7(5)9)14(12,13)8(10)11/h2-4,8H,1H3
Inchi Key PYXMDNDRKOWSMI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart