PRI424059237 Princeton BioMolecular Research, Inc.

3-(1-Methoxy-2-methyl-propyl)-phenylamine

  • MDL number MFCD20492037
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(1-methoxy-2-methylpropyl)aniline
Inchi Code 1S/C11H17NO/c1-8(2)11(13-3)9-5-4-6-10(12)7-9/h4-8,11H,12H2,1-3H3
Inchi Key NQWYAAFPZBHQSL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart