PRI424059244 Princeton BioMolecular Research, Inc.

5-Pyrrolidin-2-yl-1H-pyridin-2-one

  • MDL number MFCD20530694
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(2-pyrrolidinyl)-2(1H)-pyridinone
Inchi Code 1S/C9H12N2O/c12-9-4-3-7(6-11-9)8-2-1-5-10-8/h3-4,6,8,10H,1-2,5H2,(H,11,12)
Inchi Key CELSHJKHNJIAAJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart