PRI424059378 Princeton BioMolecular Research, Inc.

2-(6-Bromo-pyridin-3-yl)-ethanol

  • MDL number MFCD21086446
  • Cas Number : 432554-51-3
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(6-bromo-3-pyridinyl)ethanol
Inchi Code 1S/C7H8BrNO/c8-7-2-1-6(3-4-10)5-9-7/h1-2,5,10H,3-4H2
Inchi Key SDZUYYXLNCMUFA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart