PRI424059400 Princeton BioMolecular Research, Inc.

6-Amino-2-methyl-nicotinic acid methyl ester

  • MDL number MFCD08062965
  • Cas Number : 872355-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart