PRI424059401 Princeton BioMolecular Research, Inc.

(6-Amino-2-methyl-pyridin-3-yl)-methanol

  • MDL number MFCD16606248
  • Cas Number : 1227489-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart