PRI424059472 Princeton BioMolecular Research, Inc.

1-benzyl-5-methoxy-triazole-4-carboxylic acid

  • MDL number MFCD21646845
  • Cas Number : 1365940-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart