PRI424059620 Princeton BioMolecular Research, Inc.

6-amino-4,4-dimethyl-1,3-dihydroquinolin-2-one

  • MDL number MFCD11846986
  • Cas Number : 124954-76-3
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-4,4-dimethyl-3,4-dihydro-2(1H)-quinolinone
Inchi Code 1S/C11H14N2O/c1-11(2)6-10(14)13-9-4-3-7(12)5-8(9)11/h3-5H,6,12H2,1-2H3,(H,13,14)
Inchi Key NVPVNEBCSXGQHS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart