PRI424059817 Princeton BioMolecular Research, Inc.

4-Bromo-5-methyl-furan-2-carbaldehyde

  • MDL number MFCD20923653
  • Cas Number : 20627-04-7
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-methyl-2-furaldehyde
Inchi Code 1S/C6H5BrO2/c1-4-6(7)2-5(3-8)9-4/h2-3H,1H3
Inchi Key ABSGBYBACFSCEK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart