PRI424059822 Princeton BioMolecular Research, Inc.

methyl 2-chloro-6-(3-chloropropyl)pyrimidine-4-carboxylate

  • MDL number MFCD22421391
  • Cas Number : 1399653-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart