PRI424059930 Princeton BioMolecular Research, Inc.

ethyl 4-hydroxy-3-methyl-6-phenyl-benzofuran-2-carboxylate

  • MDL number MFCD02053358
  • Molecular Weight : 296.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart