PRI424059939 Princeton BioMolecular Research, Inc.

Chroman-8-ylamine

  • MDL number MFCD15527504
  • Cas Number : 113722-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart