PRI424059944 Princeton BioMolecular Research, Inc.

1-(4-Amino-phenyl)-2-methanesulfonyl-ethanol

  • MDL number MFCD22626788
  • Molecular Weight : 215.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-aminophenyl)-2-(methylsulfonyl)ethanol
Inchi Code 1S/C9H13NO3S/c1-14(12,13)6-9(11)7-2-4-8(10)5-3-7/h2-5,9,11H,6,10H2,1H3
Inchi Key IDCYNCSSGZMFHA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart