PRI424059995 Princeton BioMolecular Research, Inc.

(4-Bromo-5-methoxy-1-methyl-1H-pyrazol-3-yl)-methanol

  • MDL number MFCD26381400
  • Cas Number : 1707359-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart