PRI424060038 Princeton BioMolecular Research, Inc.

3-(2-Cyclopentyl-2H-pyrazol-3-yl)-piperidine

  • MDL number MFCD24682803
  • Cas Number : 1437432-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart