PRI424060039 Princeton BioMolecular Research, Inc.

3-(2-tert-Butyl-2H-pyrazol-3-yl)-piperidine

  • MDL number MFCD26381670
  • Cas Number : 1503966-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart