PRI424060091 Princeton BioMolecular Research, Inc.

(1-methylsulfonylcyclopropyl)methanamine

  • MDL number MFCD16766477
  • Molecular Weight : 149.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(methylsulfonyl)cyclopropyl]methanamine
Inchi Code 1S/C5H11NO2S/c1-9(7,8)5(4-6)2-3-5/h2-4,6H2,1H3
Inchi Key TZIMDZUAEJJJRT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart