PRI424060104 Princeton BioMolecular Research, Inc.

3-(2-Cyclohexyl-2H-pyrazol-3-yl)-piperidine

  • MDL number MFCD24038715
  • Cas Number : 1429901-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart