PRI424060107 Princeton BioMolecular Research, Inc.

2-[4-methoxy-3-(4-methoxyphenyl)-6-oxo-pyridazin-1-yl]propanoic acid

  • MDL number MFCD24682804
  • Cas Number : 1437432-59-1
  • Molecular Weight : 304.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-methoxy-3-(4-methoxyphenyl)-6-oxopyridazin-1(6H)-yl)propanoic acid
Inchi Code 1S/C15H16N2O5/c1-9(15(19)20)17-13(18)8-12(22-3)14(16-17)10-4-6-11(21-2)7-5-10/h4-9H,1-3H3,(H,19,20)
Inchi Key IZKJCGCPUOXDAP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart