PRI424060135 Princeton BioMolecular Research, Inc.

N-(2-Bromo-benzyl)-hydroxylamine

  • MDL number MFCD24285004
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-bromobenzyl)hydroxylamine
Inchi Code 1S/C7H8BrNO/c8-7-4-2-1-3-6(7)5-9-10/h1-4,9-10H,5H2
Inchi Key DWGTZYMYZUULRD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart