PRI424060483 Princeton BioMolecular Research, Inc.

1-[(1-methylpyrazol-4-yl)methyl]-5-nitro-pyrazole

  • MDL number MFCD04971131
  • Cas Number : 1005640-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart