PRI424060572 Princeton BioMolecular Research, Inc.

1-(2,2-Difluoro-ethyl)-4-nitro-1H-pyrazole

  • MDL number MFCD08556227
  • Cas Number : 1006442-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart